N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine

Supplier: Matrix Scientific

Synonyms: 6-Chloro-N,N-dimethylpyrazin-2-amine

051885-500MG 051885-2.5G
101897-430EA 18.46 USD
101897-430 101897-432
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine

Hazard: Irritant

Formula: C₆H₈ClN₃
MW: 157.6 g/mol
MDL Number: MFCD12024889
CAS Number: 61655-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR