1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]indoline

Supplier: Matrix Scientific

051881-500MG 051881-2.5G
101897-414EA 364.62 USD
101897-414 101897-416
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]indoline
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]indoline

Hazard: Irritant

Formula: C₁₄H₁₀ClF₃N₂
MDL Number: MFCD13561847
CAS Number: 1220027-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR