2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol

Supplier: Matrix Scientific

051879-500MG 051879-2.5G
101897-406EA 364.62 USD
101897-406 101897-408
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃N₃O
MDL Number: MFCD00203913
CAS Number: 215434-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR