N-Benzyl-3-chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

051877-500MG 051877-2.5G
101897-398EA 364.62 USD
101897-398 101897-400
N-Benzyl-3-chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine
N-Benzyl-3-chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₄H₁₂ClF₃N₂
MDL Number: MFCD13561845
CAS Number: 1220030-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR