3-Chloro-2-(2-ethyl-1-piperidinyl)-5-(trifluoromethyl)pyridine

Supplier: Matrix Scientific

051876-500MG 051876-2.5G
101897-394EA 364.62 USD
101897-394 101897-396
3-Chloro-2-(2-ethyl-1-piperidinyl)-5-(trifluoromethyl)pyridine
3-Chloro-2-(2-ethyl-1-piperidinyl)-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₃H₁₆ClF₃N₂
MDL Number: MFCD13561844
CAS Number: 1219961-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR