3-Chloro-N,N-dipropyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

051874-500MG 051874-2.5G
101897-386EA 364.62 USD
101897-386 101897-388
3-Chloro-N,N-dipropyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N,N-dipropyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₆ClF₃N₂
MDL Number: MFCD13561842
CAS Number: 1220029-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR