1-Benzyl-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine

Supplier: Matrix Scientific

051871-500MG 051871-2.5G
101897-376EA 364.62 USD
101897-376 101897-378
1-Benzyl-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
1-Benzyl-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine

Hazard: Irritant

Formula: C₁₇H₁₇ClF₃N₃
MDL Number: MFCD06243219
CAS Number: 676624-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR