1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-methylpiperazine

Supplier: Matrix Scientific

051870-500MG 051870-2.5G
101897-372EA 364.62 USD
101897-372 101897-374
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-methylpiperazine
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-methylpiperazine

Hazard: Irritant

Formula: C₁₁H₁₃ClF₃N₃
MDL Number: MFCD06243223
CAS Number: 675844-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR