3-Chloro-2-(2-methyl-1-piperidinyl)-5-(trifluoromethyl)pyridine
Supplier: Matrix Scientific
051867-500MG
051867-2.5G
101897-360EA
364.62
USD
101897-360
101897-362
3-Chloro-2-(2-methyl-1-piperidinyl)-5-(trifluoromethyl)pyridine
3-Chloro-2-(2-methyl-1-piperidinyl)-5-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₁₄ClF₃N₂ |
MDL Number:
MFCD13561838 CAS Number: 1220029-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...