N,N-Diallyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

051866-500MG 051866-2.5G
101897-356EA 364.62 USD
101897-356 101897-358
N,N-Diallyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
N,N-Diallyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₂ClF₃N₂
MDL Number: MFCD01568703
CAS Number: 478260-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR