2-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl](ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

051865-500MG 051865-2.5G
101897-352EA 364.62 USD
101897-352 101897-354
2-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl](ethyl)amino]-1-ethanol
2-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl](ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₂ClF₃N₂O
MDL Number: MFCD13223706
CAS Number: 1220033-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR