N-Butyl-3-chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

051864-500MG 051864-2.5G
101897-348EA 364.62 USD
101897-348 101897-350
N-Butyl-3-chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine
N-Butyl-3-chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₄ClF₃N₂
MDL Number: MFCD13561837
CAS Number: 1219980-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR