3-Chloro-2-(1-piperidinyl)-5-(trifluoromethyl)pyridine
Supplier: Matrix Scientific
Synonyms:
3-Chloro-2-(piperidin-1-yl)-5-(trifluoromethyl)pyridine
051861-500MG
051861-2.5G
101897-336EA
364.62
USD
101897-336
101897-338
3-Chloro-2-(1-piperidinyl)-5-(trifluoromethyl)pyridine
3-Chloro-2-(1-piperidinyl)-5-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₁₁H₁₂ClF₃N₂ MW: 264.68 g/mol |
MDL Number:
MFCD13561834 CAS Number: 1219967-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...