3-Chloro-N,N-dimethyl-5-(trifluoromethyl)-2-pyridinamine
Supplier: Matrix Scientific
Synonyms:
3-Chloro-N,N-dimethyl-5-(trifluoromethyl)pyridin-2-amine
051858-500MG
051858-2.5G
101897-324EA
364.62
USD
101897-324
101897-326
3-Chloro-N,N-dimethyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N,N-dimethyl-5-(trifluoromethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₈H₈ClF₃N₂ MW: 224.61 g/mol |
MDL Number:
MFCD13561831 CAS Number: 157071-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...