6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-pyrimidinamine

Supplier: Matrix Scientific

051857-500MG 051857-2.5G
101897-320EA 364.62 USD
101897-320 101897-322
6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-pyrimidinamine
6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O
MDL Number: MFCD13561830
CAS Number: 1220036-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR