N-Benzyl-6-chloro-N-ethyl-4-pyrimidinamine

Supplier: Matrix Scientific

051856-500MG 051856-2.5G
101897-316EA 364.62 USD
101897-316 101897-318
N-Benzyl-6-chloro-N-ethyl-4-pyrimidinamine
N-Benzyl-6-chloro-N-ethyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₃H₁₄ClN₃
MDL Number: MFCD13329208
CAS Number: 1219980-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR