1-(6-Chloro-4-pyrimidinyl)-1,2,3,4-tetrahydroquinoline

Supplier: Matrix Scientific

051855-500MG 051855-2.5G
101897-312EA 364.62 USD
101897-312 101897-314
1-(6-Chloro-4-pyrimidinyl)-1,2,3,4-tetrahydroquinoline
1-(6-Chloro-4-pyrimidinyl)-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₁₃H₁₂ClN₃
MDL Number: MFCD13329237
CAS Number: 1220020-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR