2-[4-(6-Chloro-4-pyrimidinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051852-500MG 051852-2.5G
101897-300EA 384.62 USD
101897-300 101897-302
2-[4-(6-Chloro-4-pyrimidinyl)-1-piperazinyl]-1-ethanol
2-[4-(6-Chloro-4-pyrimidinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₅ClN₄O
MDL Number: MFCD13561828
CAS Number: 1220036-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR