6-Chloro-N-cyclohexyl-N-ethyl-4-pyrimidinamine

Supplier: Matrix Scientific

051851-500MG 051851-2.5G
101897-296EA 364.62 USD
101897-296 101897-298
6-Chloro-N-cyclohexyl-N-ethyl-4-pyrimidinamine
6-Chloro-N-cyclohexyl-N-ethyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃
MDL Number: MFCD13561827
CAS Number: 1220033-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR