N-Benzyl-6-chloro-N-methyl-4-pyrimidinamine

Supplier: Matrix Scientific

051850-500MG 051850-2.5G
101897-292EA 364.62 USD
101897-292 101897-294
N-Benzyl-6-chloro-N-methyl-4-pyrimidinamine
N-Benzyl-6-chloro-N-methyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃
MDL Number: MFCD13561826
CAS Number: 1220029-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR