3-[(4-Nitrophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050785-500MG 050785-2.5G
101893-036EA 364.62 USD
101893-036 101893-038
3-[(4-Nitrophenoxy)methyl]piperidine hydrochloride
3-[(4-Nitrophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃
MDL Number: MFCD13561169
CAS Number: 1219982-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR