3-(4-Methyl-2-nitrophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050783-500MG 050783-2.5G
101893-028EA 364.62 USD
101893-028 101893-030
3-(4-Methyl-2-nitrophenoxy)pyrrolidine hydrochloride
3-(4-Methyl-2-nitrophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃
MDL Number: MFCD13561168
CAS Number: 1220037-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR