3-[(4-Methyl-2-nitrophenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
050781-500MG
050781-2.5G
101893-020EA
364.62
USD
101893-020
101893-022
3-[(4-Methyl-2-nitrophenoxy)methyl]piperidine hydrochloride
3-[(4-Methyl-2-nitrophenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₃ |
MDL Number:
MFCD13561166 CAS Number: 1220037-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...