3-(4-Methyl-2-nitrophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050779-500MG 050779-2.5G
101893-012EA 364.62 USD
101893-012 101893-014
3-(4-Methyl-2-nitrophenoxy)piperidine hydrochloride
3-(4-Methyl-2-nitrophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃
MDL Number: MFCD13561164
CAS Number: 1220034-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR