3-(4-Piperidinylmethoxy)pyridine hydrochloride
Supplier: Matrix Scientific
050777-500MG
050777-2.5G
101893-004EA
364.62
USD
101893-004
101893-006
3-(4-Piperidinylmethoxy)pyridine hydrochloride
3-(4-Piperidinylmethoxy)pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₇ClN₂O |
MDL Number:
MFCD13561162 CAS Number: 1219976-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...