3-(4-Piperidinyloxy)pyridine hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-(Piperidin-4-yloxy)-pyridine hydrochloride

050775-500MG 050775-2.5G
101892-996EA 364.62 USD
101892-996 101892-998
3-(4-Piperidinyloxy)pyridine hydrochloride
3-(4-Piperidinyloxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MW: 214.7 g/mol
MDL Number: MFCD13561160
CAS Number: 310880-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR