3-[(4-Chloro-1-naphthyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050773-500MG 050773-2.5G
101892-988EA 364.62 USD
101892-988 101892-990
3-[(4-Chloro-1-naphthyl)oxy]pyrrolidine hydrochloride
3-[(4-Chloro-1-naphthyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₅Cl₂NO
MDL Number: MFCD13561158
CAS Number: 1219976-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR