4-Chloro-1-naphthyl-4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050772-500MG 050772-2.5G
101892-984EA 364.62 USD
101892-984 101892-986
4-Chloro-1-naphthyl-4-piperidinylmethyl ether hydrochloride
4-Chloro-1-naphthyl-4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₉Cl₂NO
MDL Number: MFCD13561157
CAS Number: 1220034-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR