4-Chloro-1-naphthyl-3-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050771-500MG 050771-2.5G
101892-980EA 364.62 USD
101892-980 101892-982
4-Chloro-1-naphthyl-3-piperidinylmethyl ether hydrochloride
4-Chloro-1-naphthyl-3-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₉Cl₂NO
MW: 312.24 g/mol
MDL Number: MFCD08727956
CAS Number: 1050509-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR