4-[(4-Chloro-1-naphthyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050770-500MG 050770-2.5G
101892-976EA 364.62 USD
101892-976 101892-978
4-[(4-Chloro-1-naphthyl)oxy]piperidine hydrochloride
4-[(4-Chloro-1-naphthyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₇Cl₂NO
MDL Number: MFCD08727955
CAS Number: 1050509-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR