3-(4-Propylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050769-500MG 050769-2.5G
101892-972EA 364.62 USD
101892-972 101892-974
3-(4-Propylphenoxy)pyrrolidine hydrochloride
3-(4-Propylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13561156
CAS Number: 1220029-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR