3-[(4-Propylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050767-500MG 050767-2.5G
101892-964EA 364.62 USD
101892-964 101892-966
3-[(4-Propylphenoxy)methyl]piperidine hydrochloride
3-[(4-Propylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13561154
CAS Number: 1219963-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR