4-(4-Propylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050766-500MG 050766-2.5G
101892-960EA 364.62 USD
101892-960 101892-962
4-(4-Propylphenoxy)piperidine hydrochloride
4-(4-Propylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13561153
CAS Number: 1219976-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR