3-(4-Propylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050765-500MG 050765-2.5G
101892-956EA 364.62 USD
101892-956 101892-958
3-(4-Propylphenoxy)piperidine hydrochloride
3-(4-Propylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13561152
CAS Number: 1220034-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR