4-(tert-Pentyl)phenyl-3-pyrrolidinyl ether hydrochloride
Supplier: Matrix Scientific
050764-500MG
050764-2.5G
101892-952EA
364.62
USD
101892-952
101892-954
4-(tert-Pentyl)phenyl-3-pyrrolidinyl ether hydrochloride
4-(tert-Pentyl)phenyl-3-pyrrolidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO MW: 269.82 g/mol |
MDL Number:
MFCD13561151 CAS Number: 1219982-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...