4-(tert-Pentyl)phenyl-4-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
050761-500MG
050761-2.5G
101892-940EA
364.62
USD
101892-940
101892-942
4-(tert-Pentyl)phenyl-4-piperidinyl ether hydrochloride
4-(tert-Pentyl)phenyl-4-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13561148 CAS Number: 1219963-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...