3-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050755-500MG 050755-2.5G
101892-916EA 364.62 USD
101892-916 101892-918
3-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]piperidine hydrochloride
3-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₃₂ClNO
MDL Number: MFCD13561142
CAS Number: 1220029-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR