4-{[4-(Trifluoromethyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050752-500MG 050752-2.5G
101892-908EA 364.62 USD
101892-908 101892-910
4-{[4-(Trifluoromethyl)phenoxy]methyl}piperidine hydrochloride
4-{[4-(Trifluoromethyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClF₃NO
MDL Number: MFCD13561139
CAS Number: 1219982-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR