3-{[4-(Trifluoromethyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050751-500MG 050751-2.5G
101892-904EA 364.62 USD
101892-904 101892-906
3-{[4-(Trifluoromethyl)phenoxy]methyl}piperidine hydrochloride
3-{[4-(Trifluoromethyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClF₃NO
MDL Number: MFCD13561138
CAS Number: 1220037-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR