3-[4-(Trifluoromethyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050750-500MG 050750-2.5G
101892-900EA 364.62 USD
101892-900 101892-902
3-[4-(Trifluoromethyl)phenoxy]piperidine hydrochloride
3-[4-(Trifluoromethyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃NO
MDL Number: MFCD13561137
CAS Number: 1220033-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR