Methyl-4-(4-piperidinylmethoxy)benzoate hydrochloride
Supplier: Matrix Scientific
050748-500MG
050748-2.5G
101892-892EA
364.62
USD
101892-892
101892-894
Methyl-4-(4-piperidinylmethoxy)benzoate hydrochloride
Methyl-4-(4-piperidinylmethoxy)benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClNO₃ |
MDL Number:
MFCD13561135 CAS Number: 1219963-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...