Methyl-4-(3-piperidinylmethoxy)benzoate hydrochloride
Supplier: Matrix Scientific
050747-500MG
050747-2.5G
101892-888EA
364.62
USD
101892-888
101892-890
Methyl-4-(3-piperidinylmethoxy)benzoate hydrochloride
Methyl-4-(3-piperidinylmethoxy)benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClNO₃ |
MDL Number:
MFCD13561134 CAS Number: 1219972-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...