Methyl-4-(piperidin-4-yloxy)benzoate hydrochloride

Supplier: Matrix Scientific

Synonyms: Methyl 4-(piperidin-4-yloxy)benzoate hydrochloride

050746-500MG 050746-2.5G
101892-884EA 364.62 USD
101892-884 101892-886
Methyl-4-(piperidin-4-yloxy)benzoate hydrochloride
Methyl-4-(piperidin-4-yloxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD12546302
CAS Number: 455323-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR