Methyl-4-(3-piperidinyloxy)benzoate hydrochloride

Supplier: Matrix Scientific

050745-500MG 050745-2.5G
101892-880EA 364.62 USD
101892-880 101892-882
Methyl-4-(3-piperidinyloxy)benzoate hydrochloride
Methyl-4-(3-piperidinyloxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MW: 271.75 g/mol
MDL Number: MFCD13561133
CAS Number: 1220019-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR