3-(4-Propoxyphenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
050740-500MG
050740-2.5G
101892-860EA
364.62
USD
101892-860
101892-862
3-(4-Propoxyphenoxy)piperidine hydrochloride
3-(4-Propoxyphenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO₂ |
MDL Number:
MFCD13561128 CAS Number: 1220033-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...