N-[4-(3-Pyrrolidinyloxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

050739-500MG 050739-2.5G
101892-856EA 364.62 USD
101892-856 101892-858
N-[4-(3-Pyrrolidinyloxy)phenyl]acetamide hydrochloride
N-[4-(3-Pyrrolidinyloxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD13561127
CAS Number: 1220033-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR