N-[4-(3-Piperidinylmethoxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

050737-500MG 050737-2.5G
101892-848EA 364.62 USD
101892-848 101892-850
N-[4-(3-Piperidinylmethoxy)phenyl]acetamide hydrochloride
N-[4-(3-Piperidinylmethoxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O₂
MDL Number: MFCD13561125
CAS Number: 1220028-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR