3-(4-Iodophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050734-500MG 050734-2.5G
101892-836EA 364.62 USD
101892-836 101892-838
3-(4-Iodophenoxy)pyrrolidine hydrochloride
3-(4-Iodophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClINO
MDL Number: MFCD13561122
CAS Number: 1220019-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR