3-[(4-Iodophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050732-500MG 050732-2.5G
101892-828EA 364.62 USD
101892-828 101892-830
3-[(4-Iodophenoxy)methyl]piperidine hydrochloride
3-[(4-Iodophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClINO
MDL Number: MFCD13561120
CAS Number: 1220019-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR