3-(4-Chloro-3-methylphenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
050729-500MG
050729-2.5G
101892-816EA
166.15
USD
101892-816
101892-818
3-(4-Chloro-3-methylphenoxy)pyrrolidine hydrochloride
3-(4-Chloro-3-methylphenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅Cl₂NO |
MDL Number:
MFCD13561117 CAS Number: 1219961-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...